Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GRB7 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4856SP
This item is not returnable.
View return policy
Description
GRB7 Polyclonal specifically detects GRB7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GRB7 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B47, Epidermal growth factor receptor GRB-7, GRB7 adapter protein, growth factor receptor-bound protein 7 | |
Sheep | |
Affinity Purified | |
RUO | |
2886 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Q14451 | |
GRB7 | |
E. coli-derived recombinant human GRB7 Met130-Tyr274 Accession # Q14451 | |
25 μg | |
Primary | |
Detects endogenous human, mouse, and rat GRB7 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction