Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GRIN2A/NMDAR2A Antibody, R&D Systems™

Catalog No. PPS054
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS054 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS054 Supplier R&D Systems Supplier No. PPS054
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NMDA R, NR2A Subunit Polyclonal specifically detects NMDA R, NR2A Subunit in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NMDA R, NR2A Subunit
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias EPND, FESD, GluN1, GluN2A, LKS, MRD8, NDHMSD, NMDA R, NR2A Subunit, NMDA1, NMDAR1, NMDAR2A, NMD-R1, NR1, NR2A
Gene Symbols GRIN2A
Host Species Rabbit
Immunogen NMDA R, NR2A Subunit
Purification Method Antigen Affinity-purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2902
Test Specificity Human, mouse, and rat ∼180 kDa NMDA NR2A
Target Species Human, Mouse, Rat
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.