Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GRIN2A/NMDAR2A Antibody, R&D Systems™

Catalog No. PPS012
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS012 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS012 Supplier R&D Systems Supplier No. PPS012
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

NMDA R, NR2A Subunit Polyclonal specifically detects NMDA R, NR2A Subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen NMDA R, NR2A Subunit
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 to 1:2000 dilution, Immunoprecipitation
Gene Alias EPND, FESD, GluN1, GluN2A, LKS, MRD8, NDHMSD, NMDA R, NR2A Subunit, NMDA1, NMDAR1, NMDAR2A, NMD-R1, NR1, NR2A
Gene Symbols GRIN2A
Host Species Rabbit
Immunogen Fusion protein from the C-terminus of the NR2A subunit of rat NMDA Receptor
Purification Method Affinity Purified
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2902
Test Specificity Specific for the ∼180 kDa NR2A subunit of the NMDA Receptor in Western blots of most brain regions. No reactivity towards the NR2B and NR2C subunits. Immunolabeling is blocked by pre-adsorption of antibody with the fusion protein used to generate the antibody.
Reconstitution This antibody should be reconstituted in 50 μL phosphate buffered saline (137 mM NaCl, 7.5 mM Na2HPO4, 2.7 mM KCl, 1.5 mM KH2PO4, pH 7.4) before use.
Target Species Human, Mouse, Rat
Content And Storage After reconstitution the antibody should be aliquoted and stored at ≤ −-20°C. This product is stable at ≤ −20°C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.