Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GRIN2C/NMDAR2C Antibody, R&D Systems™

Catalog No. PPS033
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS033 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS033 Supplier R&D Systems Supplier No. PPS033
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NMDAR2C Polyclonal specifically detects NMDAR2C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NMDA R, NR2C Subunit
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunoprecipitation
Gene Alias glutamate [NMDA] receptor subunit epsilon-3, glutamate receptor, ionotropic, N-methyl D-aspartate 2C, NMDA R, NR2C Subunit, NMDAR2C, NMDAR2CNR2C, N-methyl D-aspartate receptor subtype 2C, N-methyl-D-aspartate receptor subunit 2C, NR2C
Gene Symbols GRIN2C
Host Species Rabbit
Immunogen Fusion protein from the NR2C subunit of the NMDA Receptor
Purification Method Affinity Purified
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2905
Test Specificity Human, mouse, rat ∼140 kDa NMDAR NR2C subunit
Reconstitution This antibody should be reconstituted in 50 μL phosphate buffered saline (137 mM NaCl, 7.5 mM Na 2 HPO 4 2.7 mM KCl, 1.5 mM KH 2 PO 4 pH 7.4) before use.
Target Species Human, Mouse, Rat
Content And Storage The lyophilized product is stable at < -20degrees C for at least 1 year. After reconstitution the antibody should be aliquoted and stored at < -20degrees C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.