Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Growth Hormone Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Supplier: R&D Systems AF1067
This item is not returnable.
View return policy
Description
Growth Hormone Polyclonal specifically detects Growth Hormone in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Neutralization.Specifications
Growth Hormone | |
Polyclonal | |
Unconjugated | |
CAA23779 | |
GH1 | |
E. coli-derived recombinant human Growth Hormone Phe27-Phe217 Accession # CAA23779 | |
100 μg | |
Primary | |
Detects human Growth Hormone in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 1.5-7.5 ng/mL | |
GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
Goat | |
Affinity Purified | |
RUO | |
2688 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction