Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GSK-3 beta Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC2506T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2506T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2506T100 Supplier R&D Systems Supplier No. IC2506T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen GSK-3 beta
Applications Flow Cytometry
Classification Monoclonal
Clone 272536
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P49841
Gene Alias EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform
Gene Symbols GSK3B
Host Species Rat
Immunogen E. coli-derived recombinant human GSK-3 beta Met1-Thr420 Accession # P49841
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2932
Test Specificity Detects human, mouse, and rat GSK-3 beta in ELISAs and Western blots. Does not cross-react with recombinant human GSK-3 alpha .
Target Species Human, Mouse, Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.