Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HCLS1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5789
This item is not returnable.
View return policy
Description
HCLS1 Polyclonal specifically detects HCLS1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HCLS1 | |
Polyclonal | |
Unconjugated | |
P14317 | |
HCLS1 | |
E. coli-derived recombinant human HCLS1 Met1-Gly84 Accession # P14317 | |
100 μg | |
Primary | |
Detects endogenous human, mouse and rat HCLS1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Cortactin-like, CTTNL, hematopoietic cell-specific Lyn substrate 1hematopoietic lineage cell-specific protein, HS1lckBP1, LckBP1, p75 | |
Goat | |
Affinity Purified | |
RUO | |
3059 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction