Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat HHEX Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA83771T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA83771T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA83771T100 Supplier R&D Systems Supplier No. FAB83771T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HHEX Monoclonal specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen HHEX
Applications Flow Cytometry
Classification Monoclonal
Clone 2018B
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q6UX06
Gene Alias hematopoietically expressed homeobox, Hex, HEXHMPH, HMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRH, PRHhematopoietically-expressed homeobox protein HHEX, PRHX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor
Host Species Rabbit
Immunogen E. coli-derived recombinant human HHEX Thr111-Gly270 Accession # Q6UX06
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3087
Test Specificity Detects human HHEX in direct ELISAs and Western blots.
Target Species Human, Mouse, Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.