Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat HHEX Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB83771100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB83771100 100 μg
MAB83771SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB83771100 Supplier R&D Systems Supplier No. MAB83771100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody has been used in 2 publications

HHEX Monoclonal specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen HHEX
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2018B
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 2-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. Q03014
Gene Alias hematopoietically expressed homeobox, Hex, HEXHMPH, HMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRH, PRHhematopoietically-expressed homeobox protein HHEX, PRHX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor
Host Species Rabbit
Immunogen E. coli-derived recombinant human HHEX, Thr111-Gly270, Accession # Q03014
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3087
Test Specificity Detects human HHEX in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.