Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HIF-1 alpha/HIF1A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 24 publications
Supplier: R&D Systems AF1935SP
This item is not returnable.
View return policy
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockout Validated.Specifications
HIF-1 alpha | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF 1A, HIF1, HIF1 alpha, HIF-1 alpha, HIF-1a, HIF1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
Goat | |
Affinity Purified | |
RUO | |
3091 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
ChIP Assay, Immunocytochemistry, Immunoprecipitation, KnockDown | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunoprecipitation 2 ug/500 ug cell lysate, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 0.3-15 ug/mL, Knockout Validated 0.3-15 ug/mL | |
Q16665.1 | |
HIF1A | |
E. coli-derived recombinant human HIF-1 alpha Arg575-Asn826 Accession # Q16665.1 | |
25 μg | |
Primary | |
Detects human, mouse and rat HIF-1 alpha . | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction