Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HIF-1 alpha/HIF1A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 29 publications
Supplier: R&D Systems MAB1536
This item is not returnable.
View return policy
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Knockout Validated.Specifications
HIF-1 alpha | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 1-3 ug/500 ug cell lysate, Knockout Validated | |
Q16665.1 | |
HIF1A | |
E. coli-derived recombinant human HIF-1 alpha Arg575-Asn826 Accession # Q16665.1 | |
100 μg | |
Primary | |
Detects human, mouse, and rat HIF-1 alpha . | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoassay | |
241809 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF 1A, HIF1, HIF1 alpha, HIF-1 alpha, HIF-1a, HIF1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3091 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction