Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat HIF-2 alpha/EPAS1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2997 Supplier R&D Systems Supplier No. AF2997
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. AAH57870
Gene Alias Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2
Gene Symbols EPAS1
Host Species Goat
Immunogen E. coli-derived recombinant mouse HIF-2 alpha/EPAS1 Ser542-Thr874 Accession # AAH57870
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2034
Test Specificity Detects human, mouse, and rat HIF-2 alpha/EPAS1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.