Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HOMER1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6889SP
This item is not returnable.
View return policy
Description
HOMER1 Monoclonal specifically detects HOMER1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
HOMER1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
Q86YM7 | |
HOMER1 | |
Synthetic peptide KFGQWADSRA corresponding to aa 73-82 of human HOMER1 Accession # Q86YM7 | |
25 μg | |
Primary | |
Detects human HOMER1 in direct ELISAs. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
666241 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1A, HOMER1B, HOMER-1B, HOMER1C, SYN47, SYN47neuronal immediate early gene, 1, Ves-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9456 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction