Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat HSP20/HSPB6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4200
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4200 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4200 Supplier R&D Systems Supplier No. MAB4200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody was produced using a hybridoma elicited from a mouse immunized with purified, E. coli-derived full length recombinant human Heat Shock Protein 20 kDa (rhHSP20; amino acid 1 - 160; Accession # O14558). The IgG fraction of the tissue culture supernatant was purified by protein G chromatography.

HSP20/HSPB6 Monoclonal specifically detects HSP20/HSPB6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HSP20/HSPB6
Applications Western Blot
Classification Monoclonal
Clone 494310
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and Tween™ 20 with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O14558
Gene Alias FLJ32389, Heat shock 20 kDa-like protein p20, heat shock protein beta-6, heat shock protein, alpha-crystallin-related, B6, Hsp20, HSPB6
Gene Symbols HSPB6
Host Species Mouse
Immunogen E. coli-derived recombinant human HSP20/HSPB6 Met1-Lys160 Accession # O14558
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126393
Test Specificity Detects human, mouse, and rat HSP20/HSPB6 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.