Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HSP90 beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB32861SP
This item is not returnable.
View return policy
Description
HSP90 beta Monoclonal specifically detects HSP90 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
HSP90 beta | |
Monoclonal | |
LYOPH | |
Western Blot 0.25 ug/mL, Knockout Validated | |
P08238 | |
HSP90AB1 | |
E. coli-derived recombinant human HSP90 beta Met1-Asp724 Accession # P08238 | |
25 μg | |
Primary | |
Detects human, mouse, and rat HSP90 beta in Western blots and direct ELISAs. In Western blots and direct ELISAs, no cross-reactivity with recombinant human HSP90 alpha is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoassay | |
341346 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D6S182, Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90AB1, HSP90B, HSP90-BETA, HSP90BHSP 90, HSPC2, HSPC2D6S182, HSPCB, HSPCBFLJ26984 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3326 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction