Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HTRA2/Omi Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 22 publications
Supplier: R&D Systems AF1458SP
This item is not returnable.
View return policy
Description
HTRA2/Omi Polyclonal specifically detects HTRA2/Omi in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
HTRA2/Omi | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, Omi, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
Rabbit | |
Affinity Purified | |
RUO | |
27429 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
O43464 | |
HTRA2 | |
E. coli-derived recombinant human HTRA2/Omi Ala134-Glu458 Accession # O43464 | |
25 μg | |
Primary | |
Detects human, mouse, and rat full length and mitochondria-processed HTRA2/Omi. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction