Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat IKK epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3199SP
This item is not returnable.
View return policy
Description
IKK epsilon/IKBKE Monoclonal specifically detects IKK epsilon/IKBKE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
IKK epsilon/IKBKE | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q14164 | |
IKBKE | |
E. coli-derived recombinant human IKK epsilon Met1-Val716 Accession # Q14164 | |
25 μg | |
Primary | |
Detects human, mouse, and rat IKK epsilon . | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
346909 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IKBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-I, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9641 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction