Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat ILK Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4266
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4266 Supplier R&D Systems Supplier No. MAB4266
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILK Monoclonal specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ILK
Applications Western Blot
Classification Monoclonal
Clone 443208
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13418
Gene Alias 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ESTM24, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, p59, p59ILK
Gene Symbols ILK
Host Species Mouse
Immunogen E. coli-derived recombinant human ILK Met1-Lys452 Accession # Q13418
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3611
Test Specificity Detects human, mouse, and rat ILK in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.