Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat IMP Dehydrogenase 1/IMPDH1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8554 Supplier R&D Systems Supplier No. AF8554
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IMPDH1 Polyclonal specifically detects IMPDH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen IMPDH1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Simple Western 2 μg/mL, Immunocytochemistry 1:200 dilution
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. with Sodium Azide
Gene Accession No. P20839
Gene Alias DKFZp781N0678, EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 1, IMP (inosine monophosphate) dehydrogenase 1, IMP dehydrogenase 1, IMPD, IMPD 1, IMPD1, IMPDH 1, IMPDH1, IMPDH-I, inosine-5'-monophosphate dehydrogenase 1, LCA11, LCA11RP10, retinitis pigmentosa 10 (autosomal dominant), RP10, sWSS2608
Gene Symbols IMPDH1
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf21 derived recombinaint human IMP Dehydrogenase 1/IMPDH1 Met1-Tyr563 (predicted) Accession # P20839
Purification Method Antigen Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3614
Test Specificity Detects human, rat, and mouse IMP Dehydrogenase 1/IMPDH1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse IMPDH2 is observed.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.