Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat IMP2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5305 Supplier R&D Systems Supplier No. AF5305
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IMP2/IGF2BP2 Polyclonal specifically detects IMP2/IGF2BP2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen IMP2/IGF2BP2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9Y6M1
Gene Alias Hepatocellular carcinoma autoantigen p62, IF2B2, IGF2BP2, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2
Gene Symbols IGF2BP2
Host Species Goat
Immunogen E. coli-derived recombinant human IMP2 Met1-Thr220 Accession # Q9Y6M1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10644
Test Specificity Detects endogenous human, mouse and rat IMP2 isoforms in Western blots. Reactivity with other IMP family members is unknown.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.