Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat IMPA1/IMP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB98901100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB98901-100 100 μg
MAB98901-SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB98901-100 Supplier R&D Systems Supplier No. MAB98901100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IMPA1 Monoclonal specifically detects IMPA1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen IMPA1
Applications Western Blot
Classification Monoclonal
Clone 984603
Conjugate Unconjugated
Dilution Western Blot 1μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P29218
Gene Alias EC 3.1.3.25, IMP, IMP 1, IMP1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1, MRT59, Myo-Inositol Monophosphatase 1
Host Species Mouse
Immunogen Synthetic peptide containing human IMPA/IMPA1 Peptide Accession # P29218
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3612
Test Specificity Detects human IMPA/IMPA1 Peptide in direct ELISAs. Detects human, mouse, and rat IMPA/IMPA1 in Western blots.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.