Learn More
Human/Mouse/Rat iNOS Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | iNOS |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 2D2-B2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P35228 |
| Gene Alias | EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2 |
| Gene Symbols | NOS2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.