Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Inosine 5'-Monophosphate Dehydrogenase 2/IMPDH2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8349
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8349 Supplier R&D Systems Supplier No. MAB8349
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IMP Dehydrogenase 2/IMPDH2 Monoclonal specifically detects IMP Dehydrogenase 2/IMPDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen IMP Dehydrogenase 2/IMPDH2
Applications Western Blot
Classification Monoclonal
Clone 911425
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12268
Gene Alias EC 1.1.1.205, IMDH2, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMP Oxireductase 2, IMPD 2, IMPD2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2
Gene Symbols IMPDH2
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Inosine 5'-Monophosphate Dehydrogenase 2/IMPDH2 Met1-Phe514 Accession # P12268
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3615
Test Specificity Detects human Inosine 5'-Monophosphate Dehydrogenase 2/IMPDH2 in direct ELISAs. Detects human, mouse and rat IMPDH2 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.