Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Jagged 1 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Catalog No. FAB1277T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1277T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1277T100 Supplier R&D Systems Supplier No. FAB1277T100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Jagged 1 Monoclonal specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Jagged 1
Applications Flow Cytometry
Classification Monoclonal
Clone 188331
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P78504
Gene Alias AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1
Gene Symbols JAG1
Host Species Mouse
Immunogen NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 182
Test Specificity Detects human, mouse, and rat Jagged 1. In direct ELISAs, no cross-reactivity with recombinant human Jagged 2 is observed. Stains human Jagged 1 transfectants but not the parental cell line or human Jagged 2 transfectants in flow cytometry.
Target Species Human, Mouse, Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.