Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Jagged 1 Antibody, R&D Systems™

Catalog No. MAB1277100
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1277100 100 μg
MAB1277500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1277100 Supplier R&D Systems Supplier No. MAB1277100
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

Jagged 1 Monoclonal specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Jagged 1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 188331
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P78504
Gene Alias AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1
Gene Symbols JAG1
Host Species Mouse
Immunogen NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 182
Test Specificity Detects human, mouse, and rat Jagged 1. In direct ELISAs, no cross-reactivity with recombinant human Jagged 2 is observed. Stains human Jagged 1 transfectants but not the parental cell line or human Jagged 2 transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.