Learn More
Human/Mouse/Rat Jagged 1 Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | Jagged 1 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 188331 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P78504 |
| Gene Alias | AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1 |
| Gene Symbols | JAG1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.