Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat JNK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB17761
This item is not returnable.
View return policy
Description
JNK1 Monoclonal specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
JNK1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P45983-1 | |
MAPK8 | |
E. coli-derived recombinant human JNK1 isoform 2 Ser2-Ile384 Accession # P45983-1 | |
100 μg | |
Primary | |
Detects human JNK1 in direct ELISAs and human, mouse, and rat JNK1 expressed predominantly as p46 JNK (isoforms 2 and/or 3, both 384 aa), with lesser amounts of p54 JNK (isoforms 1 and/or 4, both 427 aa), in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) JNK2 or rhJNK3 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
228601 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, MAPK8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8, PRKM8JNK, protein kinase JNK1, SAPK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5599 | |
Reconstitute at 0.5 mg/mL in sterile PBS containing 0.02% NaN 3 . | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction