Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Keap1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB3024SP
This item is not returnable.
View return policy
Description
Keap1 Monoclonal specifically detects Keap1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.Specifications
Keap1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Knockout Validated | |
Q14145 | |
KEAP1 | |
E. coli-derived recombinant human Keap1 Ala90-Ile250 Accession # Q14145 | |
25 μg | |
Primary | |
Detects human, mouse and rat Keap1 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoassay | |
333116 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9817 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction