Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Kirrel3/NEPH2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Supplier: R&D Systems AF4910SP
This item is not returnable.
View return policy
Description
Kirrel3/NEPH2 Polyclonal specifically detects Kirrel3/NEPH2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Kirrel3/NEPH2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIAA1867MGC126824, kin of IRRE like 3 (Drosophila), Kin of irregular chiasm-like protein 3, kin of IRRE-like protein 3, KIRRE, KIRREPRO4502, mKIRRE, MRD4, NEPH2, NEPH2MGC126850, nephrin-like 2, Nephrin-like protein 2 | |
Sheep | |
Affinity Purified | |
RUO | |
84623 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8IZU9 | |
KIRREL3 | |
Mouse myeloma cell line NS0-derived recombinant human Kirrel3/NEPH2 Tyr33-Ala535 Accession # Q8IZU9 | |
25 μg | |
Primary | |
Detects Kirrel3/NEPH2 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) Kirrel1 and rhKirrel2 is observed. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction