Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Kirrel3/NEPH2 Biotinylated Antibody, R&D Systems™

Catalog No. BAF4910
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4910 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4910 Supplier R&D Systems Supplier No. BAF4910
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Kirrel3/NEPH2 Polyclonal specifically detects Kirrel3/NEPH2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Kirrel3/NEPH2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot
Gene Accession No. Q8IZU9
Gene Alias KIAA1867MGC126824, kin of IRRE like 3 (Drosophila), Kin of irregular chiasm-like protein 3, kin of IRRE-like protein 3, KIRRE, KIRREPRO4502, mKIRRE, MRD4, NEPH2, NEPH2MGC126850, nephrin-like 2, Nephrin-like protein 2
Gene Symbols KIRREL3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kirrel3/NEPH2 Tyr33-Ala535 Accession # Q8IZU9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84623
Test Specificity Detects Kirrel3/NEPH2 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) Kirrel1 and rhKirrel2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.