Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Kynurenine 3-Monooxygenase/KMO Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | Kynurenine 3-Monooxygenase/KMO |
---|---|
Clone | 2493A |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8050SP
![]() |
R&D Systems
MAB8050SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8050100
![]() |
R&D Systems
MAB8050100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Kynurenine 3-Monooxygenase/KMO Monoclonal specifically detects Kynurenine 3-Monooxygenase/KMO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Kynurenine 3-Monooxygenase/KMO | |
Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
KMO, Kynurenine 3Monooxygenase, kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Kynurenine 3-Monooxygenase/KMO, Asp2-Leu441, Accession # O15229 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2493A | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O15229 | |
8564 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title