Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Lactate Dehydrogenase A/LDHA Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7304SP
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase A/LDHA Polyclonal specifically detects Lactate Dehydrogenase A/LDHA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Lactate Dehydrogenase A/LDHA | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDHA, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 | |
Sheep | |
Affinity Purified | |
RUO | |
3939 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P00338 | |
LDHA | |
E. coli-derived recombinant human Lactate Dehydrogenase A/LDHA Ala2-Val92 Accession # P00338 | |
25 μg | |
Primary | |
Detects human, mouse, and rat Lactate Dehydrogenase A/LDHA in Western blots and detects human Lactate Dehydrogenase A/LDHA in direct ELISAs. In direct ELISAs, approximately 20% cross-reactivity with recombinant human LDHB is observed. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction