Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat LAR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3004
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3004 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3004 Supplier R&D Systems Supplier No. AF3004
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

LAR/PTPRF Polyclonal specifically detects LAR/PTPRF in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen LAR/PTPRF
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. NP_569707
Gene Alias LCA-homolog, protein tyrosine phosphatase, receptor type, F, PTPRF, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Gene Symbols PTPRF
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAR Ala27-Glu1251 Accession # NP_569707
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5792
Test Specificity Detects human, mouse, and rat LAR in Western blots. In Western blots, this antibody detects both full-length protein and the cleaved LAR extracellular (E) domain. The antibody does not cross react with PTPR sigma .
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.