Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Latrophilin 3/LPHN3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5916SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5916SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5916SP Supplier R&D Systems Supplier No. MAB5916SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Latrophilin 3/LPHN3 Monoclonal specifically detects Latrophilin 3/LPHN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Latrophilin 3/LPHN3
Applications Western Blot
Classification Monoclonal
Clone 801518
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9HAR2
Gene Alias CIRL3, latrophilin 3, latrophilin homolog 3 (cow), LEC3, Lectomedin-3, LPHN3
Gene Symbols LPHN3
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Latrophilin 3/LPHN3 Met1-Arg813 Accession # Q9HAR2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23284
Test Specificity Detects human Latrophilin 3/LPHN3 in direct ELISAs and human, mouse, and rat Latrophilin 3/LPHN3 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.