Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Latrophilin 3/LPHN3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5916
This item is not returnable.
View return policy
Description
Latrophilin 3/LPHN3 Monoclonal specifically detects Latrophilin 3/LPHN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
Latrophilin 3/LPHN3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
Q9HAR2 | |
LPHN3 | |
Chinese hamster ovary cell line CHO-derived recombinant human Latrophilin 3/LPHN3 Met1-Arg813 Accession # Q9HAR2 | |
100 μg | |
Primary | |
Detects human Latrophilin 3/LPHN3 in direct ELISAs and human, mouse, and rat Latrophilin 3/LPHN3 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
801518 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CIRL3, latrophilin 3, latrophilin homolog 3 (cow), LEC3, Lectomedin-3, LPHN3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23284 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction