Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat MBP Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$227.50 - $538.00
Specifications
Antigen | MBP |
---|---|
Clone | 932908 |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 0.1-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42282SP
![]() |
R&D Systems
MAB42282SP |
25 μg |
Each for $227.50
|
|
|||||
MAB42282
![]() |
R&D Systems
MAB42282 |
100 μg |
Each for $538.00
|
|
|||||
Description
MBP Monoclonal specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
MBP | |
Western Blot 0.1 ug/mL, Immunohistochemistry 0.1-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Hmbpr, MGC99675, mld, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi | |
MBP | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
932908 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P02686 | |
4155 | |
Purified MBP from bovine brain Accession # P02686 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human, mouse, and rat MBP |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title