Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat MCHR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7938
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7938 Supplier R&D Systems Supplier No. MAB7938
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MCHR1 Monoclonal specifically detects MCHR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen MCHR1
Applications Western Blot
Classification Monoclonal
Clone 851047
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99705
Gene Alias G protein-coupled receptor 24, GPR24, GPR24SLC-1, G-protein coupled receptor 24, MCH receptor 1, MCH1R, MCH-1R, MCH1RMCHR-1, MCH-R1, melanin-concentrating hormone receptor 1, MGC32129, SLC1, SLC1MCHR, Somatostatin receptor-like protein
Gene Symbols MCHR1
Host Species Mouse
Immunogen E. coli-derived recombinant human MCHR1 Ser2-Pro113 Accession # Q99705
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2847
Test Specificity Detects human MCHR1 in ELISAs. Detects human, mouse and rat MCHR1 in Western Blots
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.