Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat MEK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2855
This item is not returnable.
View return policy
Description
MEK2 Monoclonal specifically detects MEK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
MEK2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P36507 | |
MAP2K2 | |
E. coli-derived recombinant human MEK2 Met1-Val400 Accession # P36507 | |
100 μg | |
Primary | |
Detects human, mouse, and rat MEK2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) MEK1 or rhMEK5 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry | |
300317 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ERK activator kinase 2, MAP2K2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2, MKK2FLJ26075, p45, PRKMK2MAPKK2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5605 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction