Learn More
Novus Biologicals™ Human, Mouse, Rat Methotrexate ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Description
Specifications
Specifications
| Assay Range | 0.13-1000 ng/ml |
| Assay Sensitivity | 0.087 ng/ml |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Human Serum, Mouse Serum, Plasma, Rat Serum, Urine |
| For Use With (Application) | ELISA |
| Host Species | Human, Mouse, Rat |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C |
| Target | Methotrexate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.