Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat mGluR5/1a Antibody, R&D Systems™

Catalog No. PPS079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS079 Supplier R&D Systems Supplier No. PPS079
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mGluR5/1a Polyclonal specifically detects mGluR5/1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen mGluR5/1a
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:500 dilution
Host Species Rabbit
Immunogen Peptide from the C-terminus region of rat mGluR5 and mGluR1a
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rat ∼125 kDa mGluR5 and mGluR1a monomers and ∼250 kDa homodimers
Target Species Human, Mouse, Rat
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.