Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat MKK3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2515SP
This item is not returnable.
View return policy
Description
MKK3/MEK3 Monoclonal specifically detects MKK3/MEK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| MKK3/MEK3 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MAP kinase kinase 3, MAP2K3, MAPK/ERK kinase 3, MAPKK 3, MAPKK3PRKMK3dual specificity mitogen-activated protein kinase kinase 3, MEK3, MEK3MEK 3, mitogen activated protein kinase kinase 3, mitogen-activated protein kinase kinase 3, MKK3EC 2.7.12.2, PRKMK3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5606 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 275909 | |
| Western Blot 1 ug/mL, Immunocytochemistry 15-30 ug/mL | |
| P46734 | |
| MAP2K3 | |
| E. coli-derived recombinant human MKK3 Met1-Ser347 Accession # P46734 | |
| 25 μg | |
| Primary | |
| Detects human MKK3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MKK6 is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction