Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat MKK3/MKK6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2514SP
This item is not returnable.
View return policy
Description
MKK3/MKK6 Monoclonal specifically detects MKK3/MKK6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MKK3/MKK6 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P46734 | |
E. coli-derived recombinant human MKK3 Met1-Ser347 Accession # P46734 | |
25 μg | |
Primary | |
Detects human MKK3/MKK6 in direct ELISAs and recombinant and endogenous MKK3/MKK6 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
275922 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5608 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction