Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat MKK6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF16041
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF16041 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF16041 Supplier R&D Systems Supplier No. AF16041
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MKK6/MEK6 Polyclonal specifically detects MKK6/MEK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen MKK6/MEK6
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Knockout Validated
Gene Accession No. P52564
Gene Alias MAP kinase kinase 6, MAP2K6, MAPK/ERK kinase 6, MAPKK6dual specificity mitogen-activated protein kinase kinase 6, MEK6, MEK6MAPKK 6, mitogen-activated protein kinase kinase 6, MKK6MEK 6, PRKMK6, PRKMK6protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), SAPKK3, SAPKK3EC 2.7.12.2
Gene Symbols MAP2K6
Host Species Goat
Immunogen E. coli-derived recombinant human MKK6 Ser2-Asp334 Accession # P52564
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5608
Test Specificity Detects human, mouse, and rat MKK6.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.