Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat MLK4 alpha Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF3435SP
This item is not returnable.
View return policy
Description
MLK4 alpha Polyclonal specifically detects MLK4 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MLK4 alpha | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dJ862P8.3, KIAA1804, MAP3K21, MLK4 | |
Rabbit | |
Affinity Purified | |
RUO | |
84451 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
CAC84639 | |
KIAA1804 | |
Human MLK4 alpha synthetic peptide (aa 560 - 570) ELSALPPGLLC Accession # CAC84639 | |
25 μg | |
Primary | |
Detects endogenous human, mouse, and rat MLK4 alpha . Because the peptide immunogen corresponds to a region at the C-terminus of MLK4 alpha absent in MLK4 beta , no reactivity with MLK4 beta is present. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction