Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat MST2/STK3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4549SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4549SP 25 μg
MAB4549100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4549SP Supplier R&D Systems Supplier No. MAB4549SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MST2/STK3 Monoclonal specifically detects MST2/STK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen MST2/STK3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2072A
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. Q13188
Gene Alias EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, Mess1, MST-2, MST2EC 2.7.11.1, MST3, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2, STK3
Host Species Rabbit
Immunogen E. coli-derived recombinant human STK3 Ala339-Asp456 Accession # Q13188
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6788
Test Specificity Detects human STK3 in direct ELISAs and Western blots.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.