Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat MST2/STK3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4549
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4549 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4549 Supplier R&D Systems Supplier No. AF4549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MST2/STK3 Polyclonal specifically detects MST2/STK3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MST2/STK3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q13188
Gene Alias EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, Mess1, MST-2, MST2EC 2.7.11.1, MST3, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2, STK3
Gene Symbols STK3
Host Species Goat
Immunogen E. coli-derived recombinant human MST2 Ala339-Asp456 Accession # Q13188
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6788
Test Specificity Detects endogenous human, mouse and rat MST2 in Western blots. In Western blots, this antibody shows no cross-reactivity with recombinant human MST1/STK4.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.