Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat MuRF1/TRIM63 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems AF5366SP
This item is not returnable.
View return policy
Description
MuRF1/TRIM63 Polyclonal specifically detects MuRF1/TRIM63 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
MuRF1/TRIM63 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 6.3.2.-, IRF, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZ, SMRZMURF-1, Striated muscle RING zinc finger protein, TRIM63, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63 | |
Goat | |
Affinity Purified | |
RUO | |
84676 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
Q969Q1 | |
TRIM63 | |
E. coli-derived recombinant human MuRF1/TRIM63 Met1-Gly325 Accession # Q969Q1 | |
25 μg | |
Primary | |
Detects human, mouse, and rat MuRF1/TRIM63 in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction