Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Myoglobin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$176.44 - $496.43
Specifications
| Antigen | Myoglobin |
|---|---|
| Clone | 974140 |
| Dilution | Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97201SP
![]() |
R&D Systems
MAB97201SP |
25 μg |
Each for $176.44
|
|
|||||
MAB97201100
![]() |
R&D Systems
MAB97201100 |
100 μg |
Each for $496.43
|
|
|||||
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry.Specifications
| Myoglobin | |
| Western Blot 0.05μg/mL, Simple Western 10μg/mL, ELISA, Immunohistochemistry 1-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse, Rat | |
| P02144 | |
| 4151 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 974140 | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| MB, MGC13548, myoglobin, PVALB | |
| Purified human cardiac Myoglobin antigen from human heart Accession # P02144 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Myoglobin in direct ELISAs. Detects human, mouse, and rat Myoglobin in Western blots. In sandwich immunoassays, the antibody pair is specific for human Myoglobin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title