Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat N-Cadherin Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 7 publications
Supplier: R&D Systems AF6426
This item is not returnable.
View return policy
Description
N-Cadherin Polyclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
N-Cadherin | |
Polyclonal | |
Unconjugated | |
P19022 | |
CDH2 | |
Mouse myeloma cell line NS0-derived recombinant human N-Cadherin Asp160-Ala724 Accession # P19022 | |
100 μg | |
Primary | |
Detects human, mouse, and rat N-Cadherin in Western blots. In direct ELISA, less than 10% cross-reactivity with recombinant human (rh) E-Cadherin, and rhR-Cadherin is observed. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, NCadherin, N-cadherin 1, Neural cadherin, neural-cadherin | |
Sheep | |
Affinity Purified | |
RUO | |
1000 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction