Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat NCOR1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6167SP
This item is not returnable.
View return policy
Description
NCOR1 Polyclonal specifically detects NCOR1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NCOR1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hN-CoR, KIAA1047, KIAA1047N-CoR1, MGC104216, N-CoR, N-CoRhCIT529I10, nuclear receptor corepressor 1, nuclear receptor co-repressor 1, thyroid hormone- and retinoic acid receptor-associated corepressor 1, TRAC1 | |
Sheep | |
Affinity Purified | |
RUO | |
9611 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
O75376 | |
NCOR1 | |
E. coli-derived recombinant human NCOR1 Gln1770-Ala1947 Accession # O75376 | |
25 μg | |
Primary | |
Detects human, mouse, and rat NCOR1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction